4-Bromoisoquinolin-8-amine 98%

BD304204-250MG BD304204-100MG
BLDPBD304204-250MGEA 416 EUR
BLDPBD304204-250MG BLDPBD304204-100MG
4-Bromoisoquinolin-8-amine 98%
4-Bromoisoquinolin-8-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₇BrN₂
MW: 223,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11707049
CAS Number: 351458-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR