7-Nitroisoquinolin-4-amine 95%

BD304211-1G BD304211-100MG BD304211-250MG
BLDPBD304211-1GEA 1090 EUR
BLDPBD304211-1G BLDPBD304211-100MG BLDPBD304211-250MG
7-Nitroisoquinolin-4-amine 95%
7-Nitroisoquinolin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₇N₃O₂
MW: 189,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28404726
CAS Number: 1936476-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR