3-Bromo-6,7-difluoroquinoline 95%

BD304219-250MG BD304219-1G BD304219-100MG
BLDPBD304219-250MGEA 303 EUR
BLDPBD304219-250MG BLDPBD304219-1G BLDPBD304219-100MG
3-Bromo-6,7-difluoroquinoline 95%
3-Bromo-6,7-difluoroquinoline
Formula: C₉H₄BrF₂N
MW: 244,04 g/mol
MDL Number: MFCD28404724
CAS Number: 1841081-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR