Methyl 3-aminoquinoline-6-carboxylate 95%

BD304220-100MG BD304220-1G BD304220-250MG
BLDPBD304220-100MGEA 260 EUR
BLDPBD304220-100MG BLDPBD304220-1G BLDPBD304220-250MG
Methyl 3-aminoquinoline-6-carboxylate 95%
Methyl 3-aminoquinoline-6-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₁H₁₀N₂O₂
MW: 202,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28404719
CAS Number: 1780509-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR