5-Fluoroquinolin-3-ol 95%

BD304225-100MG BD304225-1G BD304225-250MG
BLDPBD304225-100MGEA 308 EUR
BLDPBD304225-100MG BLDPBD304225-1G BLDPBD304225-250MG
5-Fluoroquinolin-3-ol 95%
5-Fluoroquinolin-3-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆FNO
MW: 163,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18414593
CAS Number: 1026265-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR