6-Aminoquinolin-3-ol 98%

BD304238-1G BD304238-250MG BD304238-100MG
BLDPBD304238-1GEA 1090 EUR
BLDPBD304238-1G BLDPBD304238-250MG BLDPBD304238-100MG
6-Aminoquinolin-3-ol 98%
6-Aminoquinolin-3-ol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₈N₂O
MW: 160,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18413399
CAS Number: 99513-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR