4-Fluoroisoquinolin-8-amine 98%

BD304249-1G BD304249-100MG BD304249-250MG
BLDPBD304249-1GEA 1730 EUR
BLDPBD304249-1G BLDPBD304249-100MG BLDPBD304249-250MG
4-Fluoroisoquinolin-8-amine 98%
4-Fluoroisoquinolin-8-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₇FN₂
MW: 162,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28975174
CAS Number: 1841081-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR