7-Fluoroisoquinolin-4-amine 95%

BD304253-1G BD304253-100MG BD304253-250MG
BLDPBD304253-1GEA 1610 EUR
BLDPBD304253-1G BLDPBD304253-100MG BLDPBD304253-250MG
7-Fluoroisoquinolin-4-amine 95%
7-Fluoroisoquinolin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₇FN₂
MW: 162,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28539788
CAS Number: 1785090-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR