3-(Trifluoromethyl)quinolin-7-ol 95%

BD304257-250MG BD304257-100MG
BLDPBD304257-250MGEA 492 EUR
BLDPBD304257-250MG BLDPBD304257-100MG
3-(Trifluoromethyl)quinolin-7-ol 95%
3-(Trifluoromethyl)quinolin-7-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₆F₃NO
MW: 213,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18417204
CAS Number: 1261471-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR