8-Fluoroisoquinolin-5-amine 98%

BD304261-250MG BD304261-1G BD304261-100MG
BLDPBD304261-250MGEA 455 EUR
BLDPBD304261-250MG BLDPBD304261-1G BLDPBD304261-100MG
8-Fluoroisoquinolin-5-amine 98%
8-Fluoroisoquinolin-5-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₇FN₂
MW: 162,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18821773
CAS Number: 608515-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR