Copper(I) diphenylphosphinate 90%

BD304293-1G BD304293-5G BD304293-25G
BLDPBD304293-1GEA 34.7 EUR
BLDPBD304293-1G BLDPBD304293-5G BLDPBD304293-25G
Copper(I) diphenylphosphinate 90%
Copper(I) diphenylphosphinate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₀CuO₂P
Storage Temperature: Ambient
MDL Number: MFCD09702021
CAS Number: 1011257-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR