5-Bromo-2-chloro-4-(difluoromethyl)pyridine 95%

BD304294-250MG BD304294-1G BD304294-25G
BLDPBD304294-250MGEA 33.6 EUR
BLDPBD304294-250MG BLDPBD304294-1G BLDPBD304294-25G
5-Bromo-2-chloro-4-(difluoromethyl)pyridine 95%
5-Bromo-2-chloro-4-(difluoromethyl)pyridine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₆H₃BrClF₂N
Storage Temperature: Freezer
MDL Number: MFCD22199288
CAS Number: 1374659-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR