3-Amino-2-methoxybenzonitrile 95%

BD304297-250MG BD304297-1G BD304297-100MG
BLDPBD304297-250MGEA 254 EUR
BLDPBD304297-250MG BLDPBD304297-1G BLDPBD304297-100MG
3-Amino-2-methoxybenzonitrile 95%
3-Amino-2-methoxybenzonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₈N₂O
MW: 148,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18389725
CAS Number: 725718-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR