4-(Bromomethyl)-2-methylthiazole hydrobromide 97%

BD304314-100MG BD304314-250MG BD304314-1G
BLDPBD304314-100MGEA 94.5 EUR
BLDPBD304314-100MG BLDPBD304314-250MG BLDPBD304314-1G
4-(Bromomethyl)-2-methylthiazole hydrobromide 97%
4-(Bromomethyl)-2-methylthiazole hydrobromide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₇Br₂NS
MW: 272,98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29044916
CAS Number: 725738-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR