trans-4-(Trifluoromethyl)cyclohexylamine 98%

Synonyms: trans-4-(Trifluoromethyl)cyclohexanamine

BLDPBD304354-250MGEA 77.7 EUR
BLDPBD304354-250MG BLDPBD304354-1G BLDPBD304354-100MG
trans-4-(Trifluoromethyl)cyclohexylamine 98%
trans-4-(Trifluoromethyl)cyclohexylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₁₂F₃N
MW: 167,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914322
CAS Number: 1073266-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR