Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%

BD304418-250MG BD304418-100MG BD304418-1G BD304418-25G BD304418-5G
BLDPBD304418-250MGEA 35.7 EUR
BLDPBD304418-250MG BLDPBD304418-100MG BLDPBD304418-1G BLDPBD304418-25G BLDPBD304418-5G
Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%
Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₂₀O₄
MW: 228,28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16036567
CAS Number: 24730-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR