Methyl 5-carbamoyl-2-methoxybenzoate 95%

BD304466-100MG BD304466-1G BD304466-50MG BD304466-250MG
BLDPBD304466-100MGEA 128 EUR
BLDPBD304466-100MG BLDPBD304466-1G BLDPBD304466-50MG BLDPBD304466-250MG
Methyl 5-carbamoyl-2-methoxybenzoate 95%
Methyl 5-carbamoyl-2-methoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₁NO₄
MW: 209,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24626283
CAS Number: 911281-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR