7-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid 95%

BD304526-250MG BD304526-100MG BD304526-1G
BLDPBD304526-250MGEA 141 EUR
BLDPBD304526-250MG BLDPBD304526-100MG BLDPBD304526-1G
7-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid 95%
7-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈N₂O₃
MW: 192,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12401189
CAS Number: 129912-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR