Molecular sieve A4 (0.4 nm, 4 Å), powder

Synonyms: Molecular sieves pack 4A,Linde 4A

BD304568-100G BD304568-1000G BD304568-25G BD304568-500G
BLDPBD304568-100GEA 7.4 EUR
BLDPBD304568-100G BLDPBD304568-1000G BLDPBD304568-25G BLDPBD304568-500G
Molecular sieve A4 (0.4 nm, 4 Å), powder
Molecular sieve A4 (0.4 nm, 4 Å)

Storage condition: Sealed in dry, Room Temperature

Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR