Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide) 95%

BD304700-250MG BD304700-1G BD304700-100MG
BLDPBD304700-250MGEA 66.2 EUR
BLDPBD304700-250MG BLDPBD304700-1G BLDPBD304700-100MG
Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide) 95%
Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₃₆H₂₆O₂P₂S
MW: 584,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27978274
CAS Number: 1019842-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR