tert-Butyl (2-(2-bromoethoxy)ethyl)carbamate 95%

BD304712-1G BD304712-250MG BD304712-5G BD304712-25G BD304712-100MG
BLDPBD304712-1GEA 86.1 EUR
BLDPBD304712-1G BLDPBD304712-250MG BLDPBD304712-5G BLDPBD304712-25G BLDPBD304712-100MG
tert-Butyl (2-(2-bromoethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-bromoethoxy)ethyl)carbamate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₉H₁₈BrNO₃
MW: 268,14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD24167474
CAS Number: 164332-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR