8-Chloro-[1,2,4]triazolo[1,5-a]pyridine 98+%

BD304910-1G BD304910-100MG BD304910-250MG
BLDPBD304910-1GEA 208 EUR
BLDPBD304910-1G BLDPBD304910-100MG BLDPBD304910-250MG
8-Chloro-[1,2,4]triazolo[1,5-a]pyridine 98+%
8-Chloro-[1,2,4]triazolo[1,5-a]pyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₄ClN₃
MW: 153,569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23708544
CAS Number: 1427368-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR