(S)-(+)-Clopidogrel sulphate 98%

Synonyms: Methyl (S)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulphate,(S)-(+)-Clopidogrel hydrogensulphate,Methyl (S)-(+)-2-(2-Chlorophenyl)-2-[6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl]acetate sulphate

BD30492-5G BD30492-25G
BLDPBD30492-5GEA 24.2 EUR
BLDPBD30492-5G BLDPBD30492-25G
(S)-(+)-Clopidogrel sulphate 98%
(S)-(+)-Clopidogrel sulphate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₆H₁₆ClNO₂S
MW: 419,89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00876395
CAS Number: 120202-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR