Bromosporine 98%

Synonyms: Ethyl (3-methyl-6-(4-methyl-3-(methylsulphonamido)phenyl)-[1,2,4]triazolo[4,3-b]pyridazin-8-yl)carbamate

BD304938-50MG
BLDPBD304938-50MGEA 693 EUR
BLDPBD304938-50MG
Bromosporine 98%
Bromosporine

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₁₇H₂₀N₆O₄S
MW: 404,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26142669
CAS Number: 1619994-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR