4-Aminoisoquinoline-8-carboxylic acid 98%

BD304982-1G BD304982-250MG BD304982-100MG
BLDPBD304982-1GEA 1380 EUR
BLDPBD304982-1G BLDPBD304982-250MG BLDPBD304982-100MG
4-Aminoisoquinoline-8-carboxylic acid 98%
4-Aminoisoquinoline-8-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₈N₂O₂
MW: 188,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28133469
CAS Number: 1822679-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR