8-Fluoroisoquinoline-4-carboxylic acid 98%

BD304985-250MG BD304985-100MG
BLDPBD304985-250MGEA 529 EUR
BLDPBD304985-250MG BLDPBD304985-100MG
8-Fluoroisoquinoline-4-carboxylic acid 98%
8-Fluoroisoquinoline-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₆FNO₂
MW: 191,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28133485
CAS Number: 1824276-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR