1,5-Dimethyl-1H-indazole-4-boronic acid 98%

Synonyms: 4-Borono-1,5-dimethyl-1H-indazole

BD305008-100MG BD305008-1G BD305008-250MG
BLDPBD305008-100MGEA 117 EUR
BLDPBD305008-100MG BLDPBD305008-1G BLDPBD305008-250MG
1,5-Dimethyl-1H-indazole-4-boronic acid 98%
1,5-Dimethyl-1H-indazole-4-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₉H₁₁BN₂O₂
MW: 190,007 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12028651
CAS Number: 1262512-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR