(S)-4-(1-Amino-2-hydroxyethyl)benzonitrile hydrochloride 95%

BLDPBD305067-1GEA 923 EUR
BLDPBD305067-1G BLDPBD305067-100MG BLDPBD305067-250MG
(S)-4-(1-Amino-2-hydroxyethyl)benzonitrile hydrochloride 95%
(S)-4-(1-Amino-2-hydroxyethyl)benzonitrile hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₁ClN₂O
MW: 198,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375653
CAS Number: 1391444-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR