2-(Methoxycarbonyl)ethylboronic acid, pinacol ester 98%

Synonyms: Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propanoate

BD305095-10G BD305095-25G
BLDPBD305095-10GEA 300 EUR
BLDPBD305095-10G BLDPBD305095-25G
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester 98%
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₉BO₄
MW: 214,066 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10700159
CAS Number: 1150561-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR