Potassium trifluoro(5-methylfuran-2-yl)borate 98%

BD305112-25G BD305112-5G BD305112-10G
BLDPBD305112-25GEA 131 EUR
BLDPBD305112-25G BLDPBD305112-5G BLDPBD305112-10G
Potassium trifluoro(5-methylfuran-2-yl)borate 98%
Potassium trifluoro(5-methylfuran-2-yl)borate

Storage condition: Sealed in dry, 2-8°C

Formula: C₅H₅BF₃KO
MW: 187,99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09992923
CAS Number: 1111213-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR