Potassium (tert-butoxymethyl)trifluoroborate 98%

Synonyms: potassium [(tert-butoxy)methyl]trifluoroboranuide,Potassium (tert-butoxymethyl)trifluoroborate,Potassium (tert-butoxymethyl)(trifluoro)borate(1-)

BD305114-5G BD305114-1G BD305114-250MG
BLDPBD305114-5GEA 156 EUR
BLDPBD305114-5G BLDPBD305114-1G BLDPBD305114-250MG
Potassium (tert-butoxymethyl)trifluoroborate 98%
Potassium (tert-butoxymethyl)trifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₁₁BF₃KO
MW: 194,046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505924
CAS Number: 910251-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR