4-Iodo-5-methyl-1H-indazole 98%

BD305213-100MG BD305213-250MG BD305213-1G BD305213-5G
BLDPBD305213-100MGEA 132 EUR
BLDPBD305213-100MG BLDPBD305213-250MG BLDPBD305213-1G BLDPBD305213-5G
4-Iodo-5-methyl-1H-indazole 98%
4-Iodo-5-methyl-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₇IN₂
MW: 258,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29056526
CAS Number: 1956309-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR