Methyl 3-oxo-3-phenylpropanoate 95%

Synonyms: Methyl 3-Oxo-3-phenylpropionate

BD305247-250MG BD305247-1G BD305247-25G BD305247-5G BD305247-100MG
BLDPBD305247-250MGEA 30.5 EUR
BLDPBD305247-250MG BLDPBD305247-1G BLDPBD305247-25G BLDPBD305247-5G BLDPBD305247-100MG
Methyl 3-oxo-3-phenylpropanoate 95%
Methyl 3-oxo-3-phenylpropanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00957180
CAS Number: 614-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR