Boc-Tyr-OtBu 98%

BD305262-1G BD305262-10G BD305262-5G BD305262-100G BD305262-500G BD305262-25G
BLDPBD305262-1GEA 7.4 EUR
BLDPBD305262-1G BLDPBD305262-10G BLDPBD305262-5G BLDPBD305262-100G BLDPBD305262-500G BLDPBD305262-25G
Boc-Tyr-OtBu 98%
Boc-Tyr-OtBu

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₈H₂₇NO₅
MW: 337,41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20264826
CAS Number: 18938-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR