7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride 95%

BD305280-250MG BD305280-100MG
BLDPBD305280-250MGEA 159 EUR
BLDPBD305280-250MG BLDPBD305280-100MG
7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride 95%
7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₄ClNOS
MW: 231,74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29056531
CAS Number: 1956386-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR