Fmoc-Trp(ME)-OH 98%

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-methyl-1H-indol-3-yl)propanoic acid

BD305288-5G BD305288-100MG BD305288-1G BD305288-10G BD305288-250MG
BLDPBD305288-5GEA 506 EUR
BLDPBD305288-5G BLDPBD305288-100MG BLDPBD305288-1G BLDPBD305288-10G BLDPBD305288-250MG
Fmoc-Trp(ME)-OH 98%
Fmoc-Trp(ME)-OH

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₇H₂₄N₂O₄
MW: 440,499 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02684512
CAS Number: 1334509-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR