(2-Fluoro-6-methoxypyridin-3-yl)boronic acid 98%

Synonyms: 2-Fluoro-6-methoxypyridine-3-boronic acid

BD305328-100MG BD305328-10G BD305328-5G BD305328-250MG BD305328-1G
BLDPBD305328-100MGEA 102 EUR
BLDPBD305328-100MG BLDPBD305328-10G BLDPBD305328-5G BLDPBD305328-250MG BLDPBD305328-1G
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid 98%
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₆H₇BFNO₃
MW: 170,93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16611244
CAS Number: 1402238-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR