2-Fluoro-N-methylpyridin-4-amine 95%

BD305347-250MG BD305347-1G BD305347-100MG
BLDPBD305347-250MGEA 64.1 EUR
BLDPBD305347-250MG BLDPBD305347-1G BLDPBD305347-100MG
2-Fluoro-N-methylpyridin-4-amine 95%
2-Fluoro-N-methylpyridin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₇FN₂
MW: 126,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18074220
CAS Number: 1564929-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR