(3,3-Difluoropiperidin-4-yl)methanol hydrochloride 98%

BD305388-100MG BD305388-250MG
BLDPBD305388-100MGEA 308 EUR
BLDPBD305388-100MG BLDPBD305388-250MG
(3,3-Difluoropiperidin-4-yl)methanol hydrochloride 98%
(3,3-Difluoropiperidin-4-yl)methanol hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₁₂ClF₂NO
MW: 187,62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27978586
CAS Number: 1783945-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR