Sodium dihydrogen phosphate hydrate 99%

Synonyms: Sodium phosphate monobasic,Sodium dihydrogen orthophosphate hydrate

BD305444-25G BD305444-1000G BD305444-100G BD305444-500G
BLDPBD305444-25GEA 6.3 EUR
BLDPBD305444-25G BLDPBD305444-1000G BLDPBD305444-100G BLDPBD305444-500G
Sodium dihydrogen phosphate hydrate 99%
Sodium dihydrogen phosphate hydrate

Storage condition: Sealed in dry, Room Temperature

Formula: NaH₂PO₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR