SGI-1027 98+%

Synonyms: N-(4-((2-amino-6-methylpyrimidin-4-yl)amino)phenyl)-4-(quinolin-4-ylamino)benzamide,N-[4-[(2-Amino-6-methyl-4-pyrimidinyl)amino]phenyl]-4-(4-quinolinylamino)benzamide

BD305455-100MG
BLDPBD305455-100MGEA 150 EUR
BLDPBD305455-100MG
SGI-1027 98+%
SGI-1027

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₇H₂₃N₇O
MW: 461,52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27937047
CAS Number: 1020149-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR