Ruthenium(III) chloride oxide 98%

Synonyms: Ruthenium oxychloride,Tetraamminechlorohydroxoruthenium(III) chloride,Ruthenium Red

BD305476-100MG BD305476-250MG BD305476-1G
BLDPBD305476-100MGEA 30.5 EUR
BLDPBD305476-100MG BLDPBD305476-250MG BLDPBD305476-1G
Ruthenium(III) chloride oxide 98%
Ruthenium(III) chloride oxide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786,354 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR