Midostaurin 99%

Synonyms: PKC412

BD305504-250MG BD305504-1G BD305504-100MG
BLDPBD305504-250MGEA 166 EUR
BLDPBD305504-250MG BLDPBD305504-1G BLDPBD305504-100MG
Midostaurin 99%
Midostaurin

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₃₅H₃₀N₄O₄
MW: 570,649 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00871372
CAS Number: 120685-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR