Hexaammineruthenium trichloride 98% Ru≥32.1%

Synonyms: Hexaammineruthenium(III) chloride,Hexaammineruthenium (III) chloride

BD305596-1G BD305596-5G BD305596-250MG
BLDPBD305596-1GEA 48.3 EUR
BLDPBD305596-1G BLDPBD305596-5G BLDPBD305596-250MG
Hexaammineruthenium trichloride 98% Ru≥32.1%
Hexaammineruthenium trichloride

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: H₁₈Cl₃N₆Ru
MW: 309,61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011478
CAS Number: 14282-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR