Methyl (5S,8S,11S)-11-(2-fluoroacetyl)-5-isopropyl-8-methyl-3,6,9-trioxo-1-phenyl-2-oxa-4,7,10-triazatridecan-13-oate 98%

BD305664-5MG BD305664-10MG BD305664-1MG
BLDPBD305664-5MGEA 226 EUR
BLDPBD305664-5MG BLDPBD305664-10MG BLDPBD305664-1MG
Methyl (5S,8S,11S)-11-(2-fluoroacetyl)-5-isopropyl-8-methyl-3,6,9-trioxo-1-phenyl-2-oxa-4,7,10-triazatridecan-13-oate 98%
Z-VAD(OMe)-FMK

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₂₂H₃₀FN₃O₇
MW: 467,48 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02684037
CAS Number: 187389-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR