4-Acetyl-L-phenylalanine Hydrochloride 95%

BLDPBD305677-1GEA 102 EUR
BLDPBD305677-1G BLDPBD305677-10G BLDPBD305677-5G BLDPBD305677-250MG BLDPBD305677-25G
4-Acetyl-L-phenylalanine Hydrochloride 95%
4-Acetyl-L-phenylalanine Hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₄ClNO₃
MW: 243,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12911919
CAS Number: 20299-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR