2,2,5,5-Tetramethyl-1,2,5-oxadisilolane 95%

BD3057-1G BD3057-5G BD3057-25G
BLDPBD3057-1GEA 27.3 EUR
BLDPBD3057-1G BLDPBD3057-5G BLDPBD3057-25G
2,2,5,5-Tetramethyl-1,2,5-oxadisilolane 95%
2,2,5,5-Tetramethyl-1,2,5-oxadisilolane

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₁₆OSi₂
MW: 160,36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040239
CAS Number: 7418-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR