TAK-715 98%

Synonyms: N-(4-(2-Ethyl-4-(m-tolyl)thiazol-5-yl)pyridin-2-yl)benzamide

BD305748-100MG BD305748-50MG BD305748-25MG
BLDPBD305748-100MGEA 449 EUR
BLDPBD305748-100MG BLDPBD305748-50MG BLDPBD305748-25MG
TAK-715 98%
TAK-715

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₄H₂₁N₃OS
MW: 399,5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17012805
CAS Number: 303162-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR