1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide 98%

BD305754-5MG
BLDPBD305754-5MGEA 110 EUR
BLDPBD305754-5MG
1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide 98%
1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₅H₁₈N₈O₅
MW: 390,35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09832544
CAS Number: 313348-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR