Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride 95%

Synonyms: Methyl 2-(S)-[N-Carbobenzyloxy]amino-3-aminopropionate, Hydrochloride

BD305772-10G BD305772-5G BD305772-1G BD305772-25G BD305772-250MG
BLDPBD305772-10GEA 376 EUR
BLDPBD305772-10G BLDPBD305772-5G BLDPBD305772-1G BLDPBD305772-25G BLDPBD305772-250MG
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride 95%
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₆N₂O₄·ClH
MW: 288,727 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701202
CAS Number: 35761-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR